
Leflunomide Related Compound B

Synonym(s):
2-Cyano-3-hydroxy-N-(4-trifluoromethylphenyl)crotonamide
Empirical Formula (Hill Notation):
C12H9F3N2O2
CAS Number:
Molecular Weight:
270.21
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2