
IAEA-S-1 (sulfur isotopes in silver sulfide)

Synonym(s):
Silver sulfide
Empirical Formula (Hill Notation):
Ag2S
CAS Number:
Molecular Weight:
247.80
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Quality Segment
Isotope
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing