
Human coagulation factor VII concentrate BRP

Synonym(s):
Factor VII from human plasma, Proconvertin
CAS Number:
Products
Manufacturer
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing