
Glimepiride Related Compound C

Synonym(s):
N-[[4-[2-[[(3-Ethyl-2,5-dihydro-4-methyl-2-oxo-1H-pyrrol-1-yl)carbonyl]amino]ethyl]phenyl]sulfonyl]carbamic acid methyl ester, Methyl ((4-(2-(3-ethyl-4-methyl-2-oxo-2,5-dihydro-1H-pyrrole-1-carboxamido)ethyl)phenyl)sulfonyl)carbamate
Empirical Formula (Hill Notation):
C18H23N3O6S
CAS Number:
Molecular Weight:
409.46
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2