
Ginsenoside Rh1

Synonym(s):
(3β,6α,12β)-3,12,20-Trihydroxydammar-24-en-6-yl-β-D-glucopyranoside, Prosapogenin A2, Sanchinoside B2, Sanchinoside Rh1
Empirical Formula (Hill Notation):
C36H62O9
CAS Number:
Molecular Weight:
638.87
Products
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing