
Fucoxanthin

Synonym(s):
all-trans-Fucoxanthin
Empirical Formula (Hill Notation):
C42H58O6
CAS Number:
Molecular Weight:
658.91
Products
Product Category
Brand
Formula Weight
Purity
Format
Research Area
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing