
Formaldehyde diethyl acetal

Synonym(s):
Diethoxymethane, Ethylal, Formaldehyde diethyl acetal
Linear Formula:
CH2(OC2H5)2
CAS Number:
Molecular Weight:
104.15
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing