
Flubendazol

Synonym(s):
Flubendazol, Flumoxanal, Methyl [5-(4-fluorobenzoyl)-1H-benzimidazol-2-yl]carbamate
Empirical Formula (Hill Notation):
C16H12FN3O3
CAS Number:
Molecular Weight:
313.28
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing