<iframe src="https://www.googletagmanager.com/ns.html?id=GTM-TXMJ2GG>m_auth=>m_preview=>m_cookies_win=x" height="0" width="0" style="display:none;visibility:hidden" id="tag-manager"></iframe>
Skip to Content
Products
Cart
0
CG
EN
Products
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Login
Back
F8BT
Synonym(s):
Poly(9,9-dioctylfluorene-
alt
-benzothiadiazole), Poly[(9,9-di-
n
-octylfluorenyl-2,7-diyl)-
alt
-(benzo[2,1,3]thiadiazol-4,8-diyl)]
Linear Formula:
(C
35
H
42
N
2
S)
n
CAS Number:
210347-52-7
Products
Brand
Sigma-Aldrich (1)
Physical Form
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
698687
F8BT,
average M
n
≤25000
Expand
View Pricing
900979
F8BT,
average M
w
>20,000
Expand
View Pricing
Choose up to 4 products to compare
Compare
Clear All
English - EN
Français - FR
Learn More