
Etomidate

Synonym(s):
(R)-1-(α-Methylbenzyl)imidazole-5-carboxylic acid ethyl ester, Amidate, R16659
Empirical Formula (Hill Notation):
C14H16N2O2
CAS Number:
Molecular Weight:
244.29
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Format
Research Area
Application
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3