
Ethyl 4-acetylbutyrate

Linear Formula:
CH3CO(CH2)3CO2C2H5
CAS Number:
Molecular Weight:
158.19
Products
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Format
Quality Segment
Markush Class
Markush Group
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing