
Ethyl 3,3-diethoxypropionate

Synonym(s):
Malonaldehydic acid ethyl ester diethylacetal
Linear Formula:
(C2H5O)2CHCH2CO2C2H5
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing