![Ethionamide European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/259/629/65c17495-0100-46d7-9874-27777e55cdc8/640/65c17495-0100-46d7-9874-27777e55cdc8.png)
Ethionamide
![Ethionamide European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/259/629/65c17495-0100-46d7-9874-27777e55cdc8/640/65c17495-0100-46d7-9874-27777e55cdc8.png)
Synonym(s):
2-Ethyl-4-pyridinecarbothioamide
Empirical Formula (Hill Notation):
C8H10N2S
CAS Number:
Molecular Weight:
166.24
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3