Skip to Content
Merck
di-Sodium hydrogen phosphate dodecahydrate crystalline, EMPROVE® ESSENTIAL, Ph. Eur., BP, ChP, JP, USP

di-Sodium hydrogen phosphate dodecahydrate

di-Sodium hydrogen phosphate dodecahydrate crystalline, EMPROVE® ESSENTIAL, Ph. Eur., BP, ChP, JP, USP
Synonym(s):
Sodium phosphate dibasic dodecahydrate, sec-Sodium phosphate, Disodium hydrogen phosphate dodecahydrate, Disodium phosphate, di-Sodium hydrogen phosphate dodecahydrate
Linear Formula:
Na2HPO4 · 12H2O
CAS Number:
Molecular Weight:
358.14

Products

Product Category
Brand
Formula Weight
Physical Form
Product Line
Agency
Chemical Composition
Quality Segment
Application