
Clofazimine

Synonym(s):
N,5-Bis(4-chlorophenyl)-3,5-dihydro-3-(isopropylimino)phenazin-2-amine
Empirical Formula (Hill Notation):
C27H22Cl2N4
CAS Number:
Molecular Weight:
473.40
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Activity Spectrum
Format
Research Area
Application
Mode of Action
API Family
Reference Material Type
Special Grade
Showing 1-4 of 4
Compare
Product Number
Description
Pricing