
CellPrime® rAlbumin

Synonym(s):
Albumin human, rHSA
Empirical Formula (Hill Notation):
C2936H4624N786O889S41
CAS Number:
Molecular Weight:
66471.53
Products
Brand
Formula Weight
Physical Form
Recombinant Host
Quality Segment
Sterility
Showing 1-1 of 1
Compare
Product Number
Description
Pricing