
Bisdemethoxycurcumin

Synonym(s):
(1E,6E)-1,7-bis(4-hydroxyphenyl)hepta-1,6-diene-3,5-dione, (E,E)-1,7-Bis(4-hydroxyphenyl)-1,6-heptadiene-3,5-dione, Bisdesmethoxycurcumin, Curcumin III, Didemethoxycurcumin
Empirical Formula (Hill Notation):
C19H16O4
CAS Number:
Molecular Weight:
308.33
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Research Area
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing