
Bakuchicin

Synonym(s):
3-(6-Hydroxy-7-benzofuranyl)-2-propenoic acid δ-lactone, 7H-Furo[2,3-f][1]benzopyran-7-one, Allopsoralen
Empirical Formula (Hill Notation):
C11H6O3
CAS Number:
Molecular Weight:
186.16
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing