
Amentoflavone

Synonym(s):
Didemethyl-ginkgetin, I3′,II8-Biapigenin, Tridemethylsciadopitysin
Empirical Formula (Hill Notation):
C30H18O10
CAS Number:
Molecular Weight:
538.46
Products
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Product Line
Purity
Format
Research Area
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing