Skip to Content
Merck
Acetyltrimethylsilane 97%

Acetyltrimethylsilane

Acetyltrimethylsilane 97%
Synonym(s):
1-(Trimethylsilyl)ethanone
Linear Formula:
(CH3)3SiCOCH3
CAS Number:
Molecular Weight:
116.23

Products

Brand
Formula Weight
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing