
Acetaldehyde ammonia trimer

Synonym(s):
Hexahydro-2,4,6-trimethyl-1,3,5-triazine trihydrate
Empirical Formula (Hill Notation):
C6H15N3 · 3H2O
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing