
7-Deaza-2′-deoxy-guanosine-5′-triphosphate

Synonym(s):
7-Deaza-2′-deoxyguanosine 5′-triphosphate lithium salt, −N7-dGTP, 7-Deaza-dGTP
Empirical Formula (Hill Notation):
C11H17N4O13P3
CAS Number:
Molecular Weight:
506.19
Products
Formula Weight
Manufacturer
Physical Form
Chemical Composition
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing