Skip to Content
Merck
6-Acetylmorphine-d6 solution 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

6-Acetylmorphine-d6 solution

6-Acetylmorphine-d6 solution 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Empirical Formula (Hill Notation):
C19D6H15NO4
CAS Number:
Molecular Weight:
333.41

Products

Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique