
5-Methyltetrahydrofolic acid disodium salt

Synonym(s):
5-Methyl-5,6,7,8-tetrahydropteroyl-L-glutamic acid disodium salt, 5-Methyl-THF, 5-Methyltetrahydrofolic acid disodium salt
Empirical Formula (Hill Notation):
C20H23N7Na2O6
CAS Number:
Molecular Weight:
503.42
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Optical Isomer
Showing 1-2 of 2