
4-Methylmorpholine N-oxide solution

Synonym(s):
NMMO solution, NMO solution, NSC 73198, NSC 82153
Empirical Formula (Hill Notation):
C5H11NO2
CAS Number:
Molecular Weight:
117.15
Products
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Quality Segment
Functional Group
Markush Class
Markush Group
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing