
4-Chlorophenylmagnesium bromide solution

Linear Formula:
ClC6H4MgBr
CAS Number:
Molecular Weight:
215.76
Products
Brand
Sustainability
Feature
Formula Weight
Physical Form
Quality Segment
Functional Group
Markush Class
Markush Group
Reaction Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing