Skip to Content
Merck
3-Maleimidobenzoic acid N-hydroxysuccinimide ester crystalline

3-Maleimidobenzoic acid N-hydroxysuccinimide ester

3-Maleimidobenzoic acid N-hydroxysuccinimide ester crystalline
Synonym(s):
MBS
Empirical Formula (Hill Notation):
C15H10N2O6
CAS Number:
Molecular Weight:
314.25

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Functional Group
Markush Class
Markush Group
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing