
2-Hexanone

Synonym(s):
Butyl methyl ketone, 2-Hexanone
Linear Formula:
CH3(CH2)3COCH3
CAS Number:
Molecular Weight:
100.16
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4