
1-Butyl-3-methylimidazolium hexafluorophosphate

Synonym(s):
BMIMPF6
Empirical Formula (Hill Notation):
C8H15F6N2P
CAS Number:
Molecular Weight:
284.18
Products
Sustainability
Feature
Formula Weight
Physical Form
Purity
Quality Segment
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing