
1,4,7-Triazacyclononane trihydrochloride

Empirical Formula (Hill Notation):
C6H15N3 · 3HCl
CAS Number:
Molecular Weight:
238.59
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing