
1,10-Phenanthroline monohydrochloride monohydrate

Synonym(s):
1,10-Phenanthroline hydrochloride monohydrate, o-Phenanthroline hydrochloride monohydrate
Empirical Formula (Hill Notation):
C12H8N2 · HCl · H2O
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing