Skip to Content
Merck
All Photos(1)

Key Documents

SMB01040

Sigma-Aldrich

23-Nor-6-oxodemethylpristimerol

≥90% (LC/MS-ELSD)

Synonym(s):

10,11-dihydroxy-2,4a,6a,6a,14a-pentamethyl-8-oxo-1,3,4,5,6,13,14,14b-octahydropicene-2-carboxylic acid, 2-Picenecarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H36O5
CAS Number:
Molecular Weight:
452.58
UNSPSC Code:
12352205
NACRES:
NA.25

biological source

plant

Assay

≥90% (LC/MS-ELSD)

form

solid

mol wt

452.58

solubility

water: slightly soluble

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

General description

2-Picenecarboxylic acid, a friedelane-type triterpenoid, is commonly found in Tripterygium hypoglaucum, Tripterygium wilfordii and Picrasma excelsa.

Application

It is a natural product derived from plant source that finds application in compound screening libraries, metabolomics, phytochemical and biochemical research.

Features and Benefits

  • High quality compound suitable for multiple research applications
  • Compatible with HPLC and mass spectrometry techniques

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service