Skip to Content
Merck
All Photos(1)

Key Documents

GE28-9034-45

GFX 96 PCR Purif Kit

Cytiva 28-9034-45, pack of 10 × 96 plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106300
NACRES:
NA.52

packaging

pack of 10 × 96 plates

manufacturer/tradename

Cytiva 28-9034-45

General description

Kit uses glass fiber matrix technology in a 96-well format for highly efficient purification of PCR products. DNA fragments from PCR are captured by the matrix in presence of a chaotropic salt and contaminants are removed by washing matrix.

Application

illustra GFX 96 Purification Kit utilizes glass fiber matrix technology in a 96-well format for highly efficient purification of PCR products. Fragments from PCR are captured by the matrix in the presence of a chaotropic salt, and contaminants are conveniently removed by washing the matrix with a buffered ethanol solution.

illustra GFX 96 PCR Purification Kit contains the following components in sufficient quantities: binding plates, wash plates, collection plates, color-coded bottles of capture buffer, wash buffer, and a choice of two elution buffers (Tris-HCl and sterile water), and an instruction booklet.

Features and Benefits

  • For the purification of up to 96 PCR products (0.1 to 10 kb) simultaneously in as little as 15 min.
  • High yields of pure DNA recovered in a small volume of water or a Low ionic strength buffer.
  • Typical recoveries are > 85% for PCR products 100 bp to 10 kb in length; salt removal typically ? 99%.
  • Prepacked glass fiber matrix 96-well plates are validated for use with Macherey-Nagel NucleoVac 96 vacuum manifold.
  • Avoids ethanol precipitations and hazardous organic extractions.
  • Purified DNA is ready for use in most applications, including fluorescent sequencing, microarrays, labeling, hybridization, ligation, and transformation.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

GFX is a trademark of Cytiva
illustra is a trademark of Cytiva

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Storage Class Code

12 - Non Combustible Liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This page shows the procedure for purifying and concentrating DNA using illustra™ GFX 96 PCR Purification Kit from Cytiva.

This page shows the procedure for purifying and concentrating DNA using illustra™ GFX 96 PCR Purification Kit from Cytiva.

This page shows the procedure for purifying and concentrating DNA using illustra™ GFX 96 PCR Purification Kit from Cytiva.

This page shows the procedure for purifying and concentrating DNA using illustra™ GFX 96 PCR Purification Kit from Cytiva.

Protocols

DNA cleanup using illustra™ GFX kits from Cytiva.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service