Key Documents
EP0030010060
Eppendorf® epT.I.P.S.® G Singles, Biopur®
volume (50-1000 μL), L (71 mm), medium blue Tray, pkg of 100 tips, individually wrapped
About This Item
UNSPSC Code:
41121600
NACRES:
NB.02
material
colorless
sterility
non-sterile
feature
individually wrapped
packaging
pack of 100 ea (individually wrapped)
pkg of 100 tips
manufacturer/tradename
Eppendorf® 0030010060
L
(71 mm)
maximum volume
1 mL
volume
(50-1000 μL)
color
medium blue Tray
Legal Information
Biopur is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
epT.I.P.S. is a registered trademark of Eppendorf AG