Skip to Content
Merck
All Photos(2)

Key Documents

CN406-05

Sigma-Aldrich

Canine Osteoblasts: CnOb (Cryovial)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NA.81

biological source

canine bone (normal)

packaging

pkg of 500,000 cells

manufacturer/tradename

Cell Applications, Inc

growth mode

Adherent

karyotype

2n = 78

morphology

osteoblast

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

arthritis; osteoporosis

shipped in

dry ice

storage temp.

−196°C

General description

Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.

Canine osteoblasts provide an excellent model system for studies relevant to the skeletal system, including mechanisms of cancerogenesis.

Canine osteoblasts have been used to define a biomarker profile of canine osteosarcoma (Milovancev, 2013), elucidate the roles of oncogenic STAT3 (Fossey, 2009, 2011) and canonical Wnt signaling (Piskun, 2013) pathways, and to develop personalized osteosarcoma treatments by testing Src inhibitor dasatinib (Davis, 2013), procaspase-3 activating compound PAC-1 (Book, 2012), HSP90 inhibitor STA-1474 (McCleese, 2009) and STAT3 inhibitor LLL12 (Couto, 2012, 2013).

Characterization: Positive for bone mineralization.

Cell Line Origin

Bone

Application

model for skeletal system research, signaling pathways, bone morphogenesis, gene expression, cell activation and proliferation, wound healing, bone formation and regeneration, treatment strategies, tissue engineering, in vitro bone models, mineralization, hormone treatments

Components

Canine Osteoblast Basal Medium containing 10% FBS & 10% DMSO

Preparation Note

  • 1st passage, >500,000 cells in Canine Osteoblast Basal Medium containing 10% FBS & 10% DMSO
  • Can be cultured at least 10 doublings

Subculture Routine

Please refer to the CnOb Culture Protocol.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service