CLS354403
Corning® BioCoat™ Fibronectin Culture Dish
diam. 60 mm
Synonym(s):
Cell Culture Dish, plastic cell culture dish, coated cell culture dish
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121812
NACRES:
NB.13
Recommended Products
feature
lid
packaging
pkg of 20 ea (4 packs )
manufacturer/tradename
Corning 354403
diam.
60 mm
binding type
Tissue Culture (TC)-treated surface
Looking for similar products? Visit Product Comparison Guide
Application
Cell culture
Legal Information
BioCoat is a trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service