Skip to Content
Merck
All Photos(1)

Key Documents

WEHI-274.1

91061231, mouse blood, Monocyte

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

WEHI-274.1, 91061231

biological source

mouse blood

growth mode

Suspension

karyotype

Not specified

morphology

Monocyte

products

Expresses the MAC-1 antigen.

receptors

Fc

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse monocyte

Cell Line Description

Derived from an Abelson virus induced tumour in a BALB/c mouse. The cells express Mac-1 but not Ia and are not phagocytic.

Culture Medium

DMEM + 2mM Glutamine + 0.05mM 2-Mercaptoethanol (2ME) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 1x100,000 to 1x1000,000 cells/ml. Adherent cells can be dislodged by scraping and re-established by centrifugation and subsequent resuspension; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service