Skip to Content
Merck
All Photos(1)

Key Documents

Huh-7D 12 Cell Line human

1042712, human liver, Epithelial

Synonym(s):

Huh7 Cells, JTC-39 Cells, Japanese Tissue Culture-39 Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

Huh-7D 12 Cell Line human, 01042712

biological source

human liver

growth mode

Adherent

karyotype

That of original Huh-7

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

storage temp.

−196°C

General description

Huh-712D is one of a series of stable HDAg (Hepatitis Delta Antigen)-expressing human hepatoma cell lines that have been isolated and characterised. Huh-7D is a derivative of Huh-7 cells, which favor hepatitis C virus replication. Huh-7 cells are superior hepatic cell models compared to HepG2 cells for apolipoprotein based metabolic studies.

Cell Line Origin

Human hepatocellular carcinoma

Cell Line Description

Huh-712D is one of a series of stable HDAg (Hepatitis Delta Antigen)-expressing human hepatoma cell lines that have been isolated and characterised.

Application

Study of intracellular traffic of delta virus antigens and RNA, viral RNA replication and early virus assembly events, and interaction between HDV and HBV.

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 11
D13S317: 10
D16S539: 10
D5S818: 12
D7S820: 11
THO1: 7
TPOX: 8,11
vWA: 16,18

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:5 i.e. seeding at 3-5x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37 C. Media change twice a week

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service