Skip to Content
Merck
All Photos(1)

Documents

Q2626

Supelco

Quinaldine phosphate salt

analytical standard

Synonym(s):

2-Methylquinoline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H9N · H3PO4
CAS Number:
Molecular Weight:
241.18
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

density

1.06 g/mL (lit.)

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

OP(O)(O)=O.Cc1ccc2ccccc2n1

InChI

1S/C10H9N.H3O4P/c1-8-6-7-9-4-2-3-5-10(9)11-8;1-5(2,3)4/h2-7H,1H3;(H3,1,2,3,4)

InChI key

RQCUDCLBKWDRRL-UHFFFAOYSA-N

Application

Quinaldine phosphate salt may be used as an analytical reference standard for the quantification of the analyte in environmental matrices using gas chromatography technique.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of trace azaarenes in water by gas chromatography and gas chromatography?mass spectrometry
Shinohara R, et al.
Journal of Chromatography A, 256, 81-91 (1983)
Characterization of nitrogen-containing aromatic compounds in soil and sediment by capillary gas chromatography-mass spectrometry after fractionation
Brumley.CW, et al.
Journal of Chromatography A, 558(1), 223-233 (1991)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service