Skip to Content
Merck
All Photos(1)

Documents

70153

Millipore

Selenite Broth (Base)

suitable for microbiology, NutriSelect® Plus

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
NACRES:
NA.74

form

powder

Quality Level

shelf life

limited shelf life, expiry date on the label

composition

bacteriological peptone, 5.0 g/L
lactose, 4.0 g/L
sodium phosphate, 10.0 g/L

manufacturer/tradename

NutriSelect® Plus

final pH

7.0±0.2 (25 °C)

application(s)

clinical testing
environmental
food and beverages

microbiology

suitability

selective for Salmonella spp.
selective for Shigella spp.

Application

A selective enrichment medium for Salmonella (including S. typhosa) from stool, urine or other pathological material.

Preparation Note

Supplement: 4 g/l of sodium hydrogen selenite (Cat. No. 71658)

Other Notes

Adaption of selenite broth for the detection of motile salmonellae

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E. Leifson
American Journal of Hygiene, 24, 423-423 (1936)
E A Hine et al.
Journal of clinical microbiology, 26(5), 875-878 (1988-05-01)
A semisolid selective motility agar based on selenite broth and designated semisolid selenite fecal (SSF) agar was developed for recovery of motile salmonellae from large numbers of rodent fecal specimens. The medium is easily prepared and inoculated; results are readily

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service