Skip to Content
Merck
All Photos(1)

Documents

62653

Millipore

LPM Agar

NutriSelect® Basic, suitable for microbiology

Synonym(s):

Listeria LPM Agar, Lithium chloride Phenylethanol Moxalactam Plating Agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171606
NACRES:
NA.74

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 15 g/L
beef extract, 3 g/L
casein peptone (pancreatic), 5 g/L
glycine anhydride, 10 g/L
lithium chloride, 5 g/L
peptone (animal), 5 g/L
phenylethyl alcohol, 2.5 g/L
sodium chloride, 5 g/L

manufacturer/tradename

NutriSelect® Basic

technique(s)

microbiological culture: suitable

final pH

7.3±0.2 (25 °C)

application(s)

agriculture
clinical testing
environmental
food and beverages

microbiology

suitability

selective for Listeria spp.

Application

For the isolation and cultivation of Listeria monocytogenes from food and dairy products.

Preparation Note

Dissolve 50.5 g in 998 ml distilled water. Heat gently and bring to the boil with constant stirring. Autoclave at 121°C for 12 minutes. Cool to 45-50°C and add aseptically 1 vial Moxalactam Supplement (Cat. No. 43963).

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service