Skip to Content
Merck
All Photos(1)

Key Documents

PMEF-NL-P1

Sigma-Aldrich

EmbryoMax® Primary Mouse Embryonic Fibroblasts

PMEF, Neo Resistant, Strain FVB, Not Treated, Passage 1

Synonym(s):

Mouse Embryonic Fibroblasts

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106509
eCl@ss:
32011203
NACRES:
NA.75

biological source

mouse

Quality Level

manufacturer/tradename

Specialty Media
EmbryoMax®

technique(s)

cell culture | stem cell: suitable

input

sample type primary embryotic fibroblasts (PMEFs)
sample type induced pluripotent stem cell(s)
sample type: mouse embryonic stem cell(s)

shipped in

liquid nitrogen

General description

PMEF-NL-P1 are low passage neomycin resistant Primary Mouse Embryo Fibroblasts (PMEFs) isolated from FVB mouse embryos at day 13. The cells have NOT been mitotically arrested. PMEFs are the traditional feeder cells used to support the growth of undifferentiated mouse and human pluripotent stem cell cultures. Additionally, low passage primary cultures of fibroblasts have a high proliferative capacity and can be used for reprogramming. It is known that cellular reprogramming efficiency declines with each passage, presumably due to lowered proliferative capacity of PMEFs at later passages. These untreated PMEF cells are high quality, low passage (P1) mouse fibroblasts that have been successfully reprogrammed into iPS cells with high efficiency. They can be used to study the mechanisms of reprogramming or be further expanded and treated with γ-irradiation or mitomycin-C to make growth arrested feeder cells. See Murine Embryonic Stem Cell Culture Manual for complete culture and media for PMEF cells.
The EmbryoMax range of PMEF cells provides researchers with a convenient solution for ES cell culture by eliminating the need for time consuming feeder cell isolation and preparation. Many embryonic stem cell culture protocols necessitate the use of primary mouse embryo fibroblast (PMEF) cells. In these protocols, ES cells are typically cultured on a monolayer of PMEF feeder cells. Feeder cells perform two important roles in stem cell culture: they secrete several important growth factors into the medium, which help maintain pluripotency, and they provide a cellular matrix for ES cells to grow.

Cell Line Description

Primary Mouse Embryo Fibroblasts

Application

Research Category
Stem Cell Research

Packaging

1x106 cells

Physical form

Provided frozen in EMD Millipore Freezing Medium (S-002-D) containing DMSO.

Storage and Stability

After receipt, vials should be stored at -80°C. If storing for longer than 6 months, store in the vapor phase of liquid nitrogen (make sure the cap is tight) for up to 2 years.

Legal Information

EmbryoMax is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Mouse embryonic fibroblasts (MEFs) serve as a feeder layer for both mouse and human embryonic stem cells (ES cells) and induced pluripotent stem cells (iPSCs).

Mouse embryonic fibroblasts (MEFs) serve as a feeder layer for both mouse and human embryonic stem cells (ES cells) and induced pluripotent stem cells (iPSCs).

Mouse embryonic fibroblasts (MEFs) serve as a feeder layer for both mouse and human embryonic stem cells (ES cells) and induced pluripotent stem cells (iPSCs).

Mouse embryonic fibroblasts (MEFs) serve as a feeder layer for both mouse and human embryonic stem cells (ES cells) and induced pluripotent stem cells (iPSCs).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service