Skip to Content
Merck
All Photos(1)

Key Documents

MABF2060

Sigma-Aldrich

Anti-IL13R alpha 1 Antibody, clone IG3

clone IG3, from mouse

Synonym(s):

Interleukin-13 receptor subunit alpha-1, IL-13 receptor subunit alpha-1, IL-13R subunit alpha-1, IL-13RA1, Interleukin-13-binding protein, Novel cytokine receptor 4, NR4, CD213a1.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

IG3, monoclonal

species reactivity

mouse

packaging

antibody small pack of 25 μg

technique(s)

flow cytometry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

mouse ... Il13Ra1(16164)

General description

Interleukin-13 receptor subunit alpha-1 (UniProt: O09030; also known as IL-13 receptor subunit alpha-1, IL-13R subunit alpha-1, IL-13R-alpha-1, IL-13RA1, Interleukin-13-binding protein, Novel cytokine receptor 4, NR4, CD213a1) is encoded by the Il13ra1 (also known as Il13r, Il13ra) gene (Gene ID: 16164) in murine species. Interleukin 13 (IL-13) is a T cell derived cytokine involved in the regulation of inflammatory and immune responses. IL-13RA1 is a single-pass type I membrane protein that binds with low affinity to interleukin-13 (IL13). Together with IL-4RA it forms a functional receptor for IL13. It is also reported to serve as an alternate accessory protein to the common cytokine receptor gamma chain for interleukin-4 (IL4) signaling, but cannot replace the function of IL2RG in allowing enhanced interleukin-2 (IL-2) binding activity. IL13-RA1 is expressed I Spleen, liver, thymus, heart, lung, kidney, testis, stomach, brain, skin, and colon. However, is not expressed in the skeletal muscle. It is also shown to bind tyrosine kinase TYK2, and thereby mediate the signaling processes that lead to the activation of JAK1, STAT3, and STAT6 induced by IL-13 and IL-4. IL-13RA1 is synthesized with a signal peptide (aa 1-25) that is cleaved off to produce mature form. It contains an extracellular domain (aa 26-340), a helical domain (aa 341-364), and a cytoplasmic region (aa 365-424). It also contain two fibronectin type III domains (aa 32-121 and 224-336). The WSXWS motif (aa 324-328) appears to be essential for proper protein folding and thereby efficient intracellular transport and cell-surface receptor binding.

Specificity

Clone IG3 detects Interleukin-13 receptor subunit alpha-1 in murine cells.

Immunogen

Recombinant mouse Interleukin-13 receptor subunit alpha-1.

Application

Anti-IL13R alpha 1, clone IG3, Cat. No. MABF2060, is a mouse monoclonal antibody that detects Interleukin-13 receptor subunit alpha-1 and has been tested for use in Flow Cytometry, Immunoprecipitation, and Western Blotting.
Flow Cytometry Analysis: A representative lot detected IL13R alpha 1 in Flow Cytometry applications (Haymaker, C.L., et. al. (2012). J Immunol. 188(7):3208-16; Dhakal, M., et. al. (2014). Eur J Immunol. 44(3):842-55).

Western Blotting Analysis: A representative lot detected IL13R alpha 1 in Western Blotting applications (Haymaker, C.L., et. al. (2012). J Immunol. 188(7):3208-16; Dhakal, M., et. al. (2014). Eur J Immunol. 44(3):842-55).

Immunoprecipitation Analysis: A representative lot detected IL13R alpha 1 in Immunoprecipitation applications (Dhakal, M., et. al. (2014). Eur J Immunol. 44(3):842-55).
Research Category
Inflammation & Immunology

Quality

Evaluated by Western Blotting in mouse spleen tissue lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected IL13R alpha 1 in 10 µg of mouse spleen tissue lysate.

Target description

~53 kDa observed; 48.40 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service