Skip to Content
Merck
All Photos(1)

Documents

AB16661

Sigma-Aldrich

Anti-PARP Poly ADP-ribose Polymerase-1 Antibody

serum, Chemicon®

Synonym(s):

Poly [ADP-ribose] polymerase 1, PARP-1, NAD(+) ADP-ribosyltransferase 1, ADPRT 1, Poly[ADP-ribose] synthase 1, ARTD1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, human, bovine, mouse

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

bovine ... Parp1(286764)
human ... PARP1(142)
mouse ... Parp1(11545)
rat ... Parp1(25591)

Specificity

Recognizes 116 kDa PARP-1 and the 85 kDa apoptosis-related cleavage fragment.

Immunogen

Synthetic peptide based on the sequence from the C-terminal portion of the bovine PARP automodification domain (amino acids 509-524) conjugated to a carrier.

Application

Research Category
Apoptosis & Cancer

Metabolism
Research Sub Category
Apoptosis - Additional

Enzymes & Biochemistry
This Anti-PARP Poly ADP-ribose Polymerase-1 Antibody is validated for use in ELISA, WB for the detection of PARP Poly ADP-ribose Polymerase-1.
Western blot: 1:500 using alkaline phosphatase.

ELISA

Optimal working dilutions must be determined by end user.

Linkage

Replaces: 04-575; 04-576

Physical form

Antiserum containing 0.05% sodium azide. Liquid containing 0.05% sodium azide.

Storage and Stability

Maintain frozen at -20°C in undiluted aliquots for up to 12 months.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lauren K Jillson et al.
Molecular cancer research : MCR, 19(7), 1123-1136 (2021-04-14)
Prostate cancer genomic subtypes that stratify aggressive disease and inform treatment decisions at the primary stage are currently limited. Previously, we functionally validated an aggressive subtype present in 15% of prostate cancer characterized by dual deletion of MAP3K7 and CHD1.
Kwang Il Park et al.
Oncology letters, 13(4), 2521-2530 (2017-04-30)
Lonicera japonica Thunb. (L. japonica T.) has historically been used in Korean herbal medicine due to its anticancer and protective effects on the respiratory system. In the present study, the polyphenolic compounds in L. japonica T. were investigated using high-performance
Hypermethylated in cancer 1 (HIC1), a tumor suppressor gene epigenetically deregulated in hyperparathyroid tumors by histone H3 lysine modification.
Svedlund, J; Koskinen Edblom, S; Marquez, VE; ?kerstrom, G; Bjorklund, P; Westin, G
The Journal of clinical endocrinology and metabolism null
Marlena Zyśk et al.
International journal of molecular sciences, 21(22) (2020-11-18)
The N-acetylaspartate network begins in neurons with N-acetylaspartate production catalyzed by aspartate N-acetyltransferase from acetyl-CoA and aspartate. Clinical studies reported a significant depletion in N-acetylaspartate brain level in type 1 diabetic patients. The main goal of this study was to
Jessica Svedlund et al.
Endocrine-related cancer, 21(2), 231-239 (2013-12-03)
Primary hyperparathyroidism (pHPT) resulting from parathyroid tumors is a common endocrine disorder with incompletely understood etiology. In renal failure, secondary hyperparathyroidism (sHPT) occurs with multiple tumor development as a result of calcium and vitamin D regulatory disturbance. The aim of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service