Skip to Content
Merck
All Photos(1)

Key Documents

8.14219

Sigma-Aldrich

N-tert-Butyldimethylsilyl-N-methyltrifluoroacetamide

for synthesis

Synonym(s):

N-tert-Butyldimethylsilyl-N-methyltrifluoroacetamide, MTBSTFA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H18F3NOSi
CAS Number:
Molecular Weight:
241.33
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.22

Quality Level

Assay

≥97.0% (GC)

form

liquid

transition temp

flash point 52 °C

density

1.03 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C9H18F3NOSi/c1-5-6-7-15(3,4)13(2)8(14)9(10,11)12/h5-7H2,1-4H3

InChI key

NEJIKZHUEMMAEF-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.034 - 1.039
Identity (IR): passes test

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

113.0 °F - closed cup

Flash Point(C)

45 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service