Skip to Content
Merck
All Photos(1)

Key Documents

441201-M

Millipore

MACERASE Pectinase, Rhizopus sp.

Useful in isolating intact cells and protoplasts from higher plants. Note: 1 KU = 1000 units.

Synonym(s):

MACERASE Pectinase, Rhizopus sp.

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.54

form

solid

specific activity

≥3000 units/g dry wt.

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

pale amber

solubility

water: 1 mg/mL

storage temp.

2-8°C

General description

Useful in isolating intact cells and protoplasts from higher plants.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is a measure of the macerating activity on potato tuber disks as determined by loss of weight at 40°C, pH 5.0.

Other Notes

Shi, H., et al. 2003. Plant Cell15, 19.
Gomez-Maldonado, J., et al. 2001. Plant Mol. Biol. Reporter 19, 361.
Takebe, I. 1968. Plant Cell Physiol.9, 115.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service