2430-M
L-Cysteine, Hydrochloride
Synonym(s):
L-Cysteine, Hydrochloride, Cys, HCI
About This Item
Recommended Products
Assay
>98% (titration)
Quality Level
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
color
white
solubility
water: 10 mg/mL
ethanol: soluble
storage temp.
10-30°C
SMILES string
SC[C@H]([N+H3])C(=O)O.[Cl-]
InChI
1S/C3H7NO2S.ClH/c4-2(1-7)3(5)6;/h2,7H,1,4H2,(H,5,6);1H/t2-;/m0./s1
InChI key
IFQSXNOEEPCSLW-DKWTVANSSA-N
General description
Application
- Application of L-Cysteine Hydrochloride Delays the Ripening of Harvested Tomato Fruit.: This research demonstrates the effectiveness of L-cysteine hydrochloride in delaying the ripening process in harvested tomatoes, suggesting a potential approach for extending the shelf life of fresh produce and reducing post-harvest losses (Song et al., 2024).
- L-Cysteine hydrochloride inhibits Aspergillus flavus growth and AFB(1) synthesis by disrupting cell structure and antioxidant system balance.: Highlights the antimicrobial properties of L-cysteine hydrochloride against Aspergillus flavus, presenting a promising compound for controlling fungal contamination in food products and enhancing food safety (Qiu et al., 2023).
Warning
Reconstitution
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service