Skip to Content
Merck
All Photos(1)

Key Documents

1.50167

Supelco

Purospher® RP-18 endcapped (5 µm) HPLC Columns

L × I.D. 4 mm x 4 mm LiChroCART Guard-cartridge

Synonym(s):

Purospher® RP-18 endcapped (5µm) LiChroCART 4-4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115709
NACRES:
SB.52

product name

Purospher® RP-18 endcapped (5 µm), L × I.D. 4 mm x 4 mm LiChroCART Guard-cartridge

material

stainless steel column

Quality Level

Agency

suitable for USP L1

product line

Purospher®

feature

endcapped

packaging

10 pak of

extent of labeling

18% Carbon loading

parameter

250 bar pressure
65 °C max. temp.

technique(s)

HPLC: suitable
LC/MS: suitable
mass spectrometry (MS): suitable (High)

column L × I.D.

0.4 cm × 4 mm

surface area

350 m2/g

matrix

fully porous particle

matrix active group

C18 bonding phase

particle size

5 μm

pore size

120 Å pore size

operating pH

2-7.5

separation technique

reversed phase

storage temp.

no temp limit

General description

HPLC guard cartidge

Application


  • Determination of nifedipine in serum of women in preterm labor by high-performance liquid chromatography with diode array detection.: This study utilized Purospher® RP-18 endcapped (5 µm) columns in the HPLC analysis of nifedipine levels in serum samples from women in preterm labor. The research demonstrated the effectiveness of the Purospher® RP-18 columns in achieving high resolution and sensitivity, making it a valuable tool for bioanalytical applications in clinical chemistry (Kowalczuk et al., 2006).

Legal Information

LICHROCART is a trademark of Merck KGaA, Darmstadt, Germany
Purospher is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service