Skip to Content
Merck
All Photos(1)

Key Documents

860681P

Avanti

C15 Ceramide-d7 (d18:1-d7/15:0)

Avanti Research - A Croda Brand 860681P, powder

Synonym(s):

N-pentadecanoyl-D-erythro-sphingosine(d7)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H58 D7NO3
CAS Number:
Molecular Weight:
530.92
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.25
Pricing and availability is not currently available.

form

powder

packaging

pkg of 1 × 1 mg (860681P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860681P

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC[C@]([H])(NC(CCCCCCCCCCCCCC)=O)[C@]([H])(O)/C=C/CCCCCCCCCCC([2H])([2H])C([2H])([2H])C([2H])([2H])[2H]

InChI key

HBULQAPKKLNTLT-CVNQHAACSA-N

General description

C15 Ceramide (d18:1/15:0) is a synthetic ceramide containing sphingosine with pentadecanoic acid.[1] C15 Ceramide-d7 (d18:1-d7/15:0) is a deuterated derivative of C15 Ceramide (d18:1/15:0).

Application

C15 Ceramide-d7 (d18:1-d7/15:0) has been used as a standard to measure C32–C34 ceramides in zebrafish brain and human embryonic kidney cell line (HEK293) by thermo q-exactive plus quadrupole-orbitrap mass spectrometery.[2] It has also been used to measure the concentration of sphingolipids in right submandibular glands of mice by ultra-high-performance liquid chromatography-tandem mass spectrometry (UHPLC/MS/MS).[3]

Biochem/physiol Actions

Ceramides serve as precursors for several sphingolipids. Ceramides play a significant role in cell differentiation, proliferation, and apoptosis. In addition, they also acts as secondary transmitters in cell signaling.[3]

Packaging

5 mL Amber Glass Screw Cap Vial (860681P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Claude Vieu et al.
Journal of lipid research, 43(3), 510-522 (2002-03-15)
This study reports a single-step analysis of the molecular species of endogenous ceramides and of the ceramide moiety of sphingomyelins in biological samples, using gas liquid chromatography (GLC). Silylated sphingomyelins were quantitatively converted to monosilylated ceramide upon injection into GLC
Anna Zalewska et al.
Biomolecules, 9(12) (2019-12-19)
This is the first study to investigate the relationship between ceramides, the mitochondrial respiratory system, oxidative stress, inflammation, and apoptosis in the submandibular gland mitochondria of mice with insulin resistance (IR). The experiment was conducted on 20 male C57BL/6 mice
Tejia Zhang et al.
Scientific reports, 9(1), 19939-19939 (2019-12-29)
Extensive characterisations of the zebrafish genome and proteome have established a foundation for the use of the zebrafish as a model organism; however, characterisation of the zebrafish lipidome has not been as comprehensive. In an effort to expand current knowledge
Hady Razak Hady et al.
Journal of clinical medicine, 8(12) (2019-12-18)
The liver plays a central role in the glucose and lipid metabolism. Studies performed on animal models have shown an important role of lipid accumulation in the induction of insulin resistance. We sought to explain whether in obese humans, the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service