Skip to Content
Merck
All Photos(1)

Key Documents

810103C

Avanti

16:0-LR/18:1/18:1 TG - lissamine rhodamine

Avanti Research - A Croda Brand 810103C

Synonym(s):

1,2-dioleoyl-3-[16-N-(lissamine rhodamine B sulfonyl) amino]palmitoyl-sn-glycerol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C82H13N3O12S2
CAS Number:
Molecular Weight:
1296.12
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (810103C-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810103C

concentration

1 mg/mL (810103C-1mg)

shipped in

dry ice

storage temp.

−20°C

General description

Lissamine rhodamine is a red-emitting fluorophore, mainly used in immunofluorescence studies. 16:0-LR/18:1/18:1 TG - lissamine rhodamine weighs around 119.38 g/mol and is available as a colourless liquid.

Packaging

5 mL Amber Glass Screw Cap Vial (810103C-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Which fluorophore is brightest? A comparison of the staining obtained using fluorescein, tetramethylrhodamine, lissamine rhodamine, Texas red, and cyanine 3.18
Wessendorf MW and Brelje TC
Histochemistry (1992)
Which fluorophore is brightest? A comparison of the staining obtained using fluorescein, tetramethylrhodamine, lissamine rhodamine, Texas red, and cyanine 3.18
Wessendorf MW and Brelje TC
Histochemistry (1992)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service